Order Entry
Belgium
ContactUsLinkComponent
Neopor MonoBox
Neopor MonoBox
  DLTTB100008
 :  DELTA T
Neopor MonoBox
  DLTTB100008
 :  DELTA T

 

  • Colour:
    Grey
  • Description:
    Insulation: 60 - 80 mm, 0,76 kg
  • Pk:
    1

 

 

Neopor packaging for 1 pair of TempShell elements. Consists of two equal halves.

  • External dimensions: 390×290×230 mm
  • Internal dimensions: 270×160×60 mm