L-Phenylalanine ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:63-91-2
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00064227
:165.19 g/mol
Merck Index:14,07271
Boiling Pt:313 °C (1013 hPa)
Melting Pt:270…275 °C (lit.)
Density:0,754 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-568-1
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- UV/VIS: l max (water):257 ± 3 nm 257.3 nm
- Optical Rotation (c=2, water):-35° ± 4°
Specification Test Results
Frequently Bought Together