L-Phenylalanine
Catalog # EDQMP1150000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:60 mg
- Seq:2385
Specifications
About this item
:
:165.19 g/mol
Merck Index:14,07271
Boiling Pt:313 °C (1013 hPa)
Melting Pt:173 °C
Density:0.754 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-568-1
CAS Number:63-91-2
Formula:C₉H₁₁NO₂
MDL Number:MFCD00064227