Order Entry
Belgium
Orders LinkContactUsLinkComponent
Assistent® Drinking Straw Cleaning Brushes

Assistent® Drinking Straw Cleaning Brushes

Nylon, 10 mm, Steel wire, 250 mm
Catalog # HECH44035010
Assistent® Drinking Straw Cleaning Brushes
Catalog # HECH44035010
Assistent® Drinking Straw Cleaning Brushes
Assistent® Drinking Straw Cleaning Brushes

Nylon, 10 mm, Steel wire, 250 mm

Price
3,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Brushes for cleaning drinking straws feature steel wire handles and nylon bristles.

Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    10 mm
  • Handle material:
    Steel wire
  • Seq:
    01
  • Total length:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.