Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Phosphate Buffer Mixed pH 4,0

Phosphate buffer mixed, 4,0, Phosphate, 1 L
  ACSDBP556
 :   CPACHEM
Phosphate Buffer Mixed pH 4,0
  ACSDBP556
 :  CPACHEM
 :  
Phosphate Buffer Mixed pH 4,0

Phosphate buffer mixed, 4,0, Phosphate, 1 L

 :  
 
108,00 €
   
1 * 1 L
 

Shelf Life on Ship Date: 12 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

 : ISO 9001.

 

 

  • pH buffer type:
    Phosphate
  • Pk:
    1 L
  • Description:
    Phosphate buffer mixed
  • pH value:
    4,0

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.