

Meloxicam
Catalog # PROOTRCM216100-250
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Pale yellow to light yellow solid
- NMR:Conforms
- Elemental analysis:%C: 47.69, %H: 3.58, %N: 11.73
- HPLC purity:99.88% (355 nm)
- MS:Conforms
Specification Test Results
About this item
:
:351.40 g/mol
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:71125-38-7
Formula:C₁₄H₁₃N₃O₄S₂
MDL Number:MFCD00868752