ContactUsLinkComponent
96-Well Autosampler Vial Plate, with Inserts
96-Well Autosampler Vial Plate, with Inserts
Catalog # MICS95096-710-D
Supplier:  MICROSOLV
undefined
96-Well Autosampler Vial Plate, with Inserts
Catalog # MICS95096-710-D
Supplier:  MICROSOLV
Specifications
  • Description:
    96 well plate with 1 ml glass inserts
  • Pk:
    250
Specifications
About this item
96 well plates with glass inserts.
  • High grade, pure silicone rubber with PTFE coating
  • Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass
  • Chemically inert
The inserts are removable and the plates can be reused.