Orders LinkContactUsLinkComponent
Hightextra® Smooth Wipes
Hightextra® Smooth Wipes
Smooth wipes, 68 gsm, White
Catalog # ECOLHTS0060R05300W
Supplier:   ECOLINE NV SA
Hightextra® Smooth Wipes
Catalog # ECOLHTS0060R05300W
Supplier:  ECOLINE NV SA
Hightextra® Smooth Wipes
Smooth wipes, 68 gsm, White
Price
77,40 €
Log in for availability
1 * 5 ROU
Product Details & Documents
The combination of woodpulp and continuous polyester fibres creates a super strong wipe, even when used wet, that copes with tougher cleaning too.
Specifications
  • Colour:
    White
  • Description:
    Smooth wipes, 68 gsm