Order Entry
Belgium
ContactUsLinkComponent
Yttrium Standard Solution, 1000 mg/L Y in Diluted Nitric Acid for ICP-MS
Yttrium Standard Solution, 1000 mg/L Y in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM267.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Yttrium Standard Solution, 1000 mg/L Y in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM267.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item