Indium Standard Solution, 1000 mg/L In in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM226.2NP.L1
Supplier: CPACHEM
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Indium
- Symbol:In