Order Entry
Belgium
ContactUsLinkComponent
Germanium Standard Solution, 1000 mg/L Ge in Diluted HNO₃ and HF for ICP-MS
Germanium Standard Solution, 1000 mg/L Ge in Diluted HNO₃ and HF for ICP-MS
Catalog # ACSDM222.2N2FP.L1
Supplier:  CPACHEM
CAS Number:  
Germanium Standard Solution, 1000 mg/L Ge in Diluted HNO₃ and HF for ICP-MS
Catalog # ACSDM222.2N2FP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃/2% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Germanium
  • Symbol:
    Ge

Specifications

About this item