Order Entry
Belgium
ContactUsLinkComponent
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Catalog # ACSDM208.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid Standard for ICP-MS
Catalog # ACSDM208.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Bi 1000 mg/L in diluted nitric acid for ICP/MS.

Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.