Order Entry
Belgium
ContactUsLinkComponent
Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid Standard for ICP-MS
Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid Standard for ICP-MS
Catalog # ACSDM132.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid Standard for ICP-MS
Catalog # ACSDM132.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Mg 10000 mg/L in diluted nitric acid for ICP/MS.

Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Certifications: ISO 17034, ISO 17025, ISO 9001.