Order Entry
Belgium
ContactUsLinkComponent
Anti-DHX37 Rabbit Polyclonal Antibody
Anti-DHX37 Rabbit Polyclonal Antibody
Catalog # NOVUNBP3-18056
Supplier:  Novus Biologicals
Anti-DHX37 Rabbit Polyclonal Antibody
Catalog # NOVUNBP3-18056
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DHX37
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    probable ATP-dependent RNA helicase DHX37|MGC2695|MGC4322|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37|DEAH (Asp-Glu-Ala-His) box polypeptide 37|KIAA1517MGC46245|DDX37|DEAH box protein 37|EC 3.6.4.13|FLJ41974|EC 3.6.1|DEAD/DEAH box helicase DDX37
  • UniProtKB:
    Q8IY370
  • Purity:
    Immunogen affinity purified
  • Storage buffer:
    PBS, contains 0,02% sodium azide
  • Storage temperature:
    Store at 4 °C short term, aliquot and store at −20 °C long term, avoid freeze-thaw cycles
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    Synthetic peptide made to an internal portion of human DHX37 (between amino acids 650-700)
  • Tested applications:
    Immunocytochemistry/ Immunofluorescence 1 - 2 µg/ml
    Immunohistochemistry 1:200
    Immunohistochemistry-Paraffin 1:200
    Western Blot 1 - 2 µg/ml
  • Pk:
    0,1 mg

Specifications

About this item

DHX37 is a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases.

They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.This gene encodes a DEAD box protein. DEAD box proteins, characterised by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases.

They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.

Type: Primary
Antigen: DHX37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse