Order Entry
Belgium
ContactUsLinkComponent
EPA Method 525.3 OCP Cal Standard, 500 µg/ml, Acetone, 1 ml/Ampul
EPA Method 525.3 OCP Cal Standard, 500 µg/ml, Acetone, 1 ml/Ampul
Catalog # REST32542
Supplier:  Restek
CAS Number:  
EPA Method 525.3 OCP Cal Standard, 500 µg/ml, Acetone, 1 ml/Ampul
Catalog # REST32542
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml
  • Pk:
    1
  • Description:
    EPA method 525.3 OCP cal standard, acetone solvent, 1 ml
  • Shipping conditions:
    Ambient
  • Storage temperature:
    10 °C or colder

Specifications

About this item

Developed specifically for EPA Method 525.3.

  • Formulated to provide maximum stability in concentrations convenient for dilution
  • Simplifies and speeds up standard preparation
  • Quantitatively tested to confirm composition; detailed support documentation provided

29 compounds:
Acetochlor (34256-82-1)
Aldrin (309-00-2)
cis-Chlordane (5103-71-9)
trans-Chlordane (5103-74-2)
Chlorobenzilate (510-15-6)
Chloroneb (2675-77-6)
Chlorothalonil (1897-45-6)
Dacthal (DCPA) (1861-32-1)
4,4'-DDD (72-54-8)
4,4'-DDE (72-55-9)
4,4'-DDT (50-29-3)
Dieldrin (60-57-1)
Endosulfan I (959-98-8)
Endosulfan II (33213-65-9)
Endosulfan sulfate (1031-07-8)
Endrin (72-20-8)
α-HCH (319-84-6)
β-HCH (319-85-7)
γ-HCH (Lindane) (58-89-9)
δ-HCH (319-86-8)
Heptachlor (76-44-8)
Heptachlor epoxide (1024-57-3)
Hexachlorobenzene (118-74-1)
Hexachlorocyclopentadiene (HCCPD) (77-47-4)
Methoxychlor (72-43-5)
trans-Nonachlor (39765-80-5)
Pentachlorophenol (87-86-5)
cis-Permethrin (61949-76-6)
trans-Permethrin (61949-77-7)

Shipping weight: 0,022 kg
L×W×H: 90×31,75×31,75 mm