Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tyramine

Tyramine

100 mg
Catalog # CMPLSB49364.100MG
Tyramine
Catalog # CMPLSB49364.100MG
CAS Number:  51-67-2
Tyramine

100 mg

CAS Number:  51-67-2
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents

Certifications: ISO 17034, ISO 17025, ISO 9001.

Ordering information: Store in a refrigerator at temperatures between 2 to 8 °C.

Synonyms:  4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
  • CAS Number:51-67-2
  • Formula:C₈H₁₁NO
  • MDL Number:MFCD00008193
  • Molecular Weight:137.18 g/mol
Specifications

Specifications

  • Pk:
    100 mg

Specifications