Pharmaceutical Analytical Standard, Low-substituted Hydroxypropylcellulose, Reference Material
Low-substituted hydroxypropylcellulose, Analytical reference standard, 50 mg
Catalog # EDQMY0001798
Supplier: EDQM
Pharmaceutical Analytical Standard, Low-substituted Hydroxypropylcellulose, Reference Material
Catalog # EDQMY0001798
Supplier: EDQM
CAS Number:
Jump to:
Product Details & Documents
Low-substituted hydroxypropylcellulose EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Specifications
Specifications
- Pk:50 mg
- Standards:Analytical reference standard
- Description:Low-substituted hydroxypropylcellulose
- Cat. No.:EDQMY0001798
- Storage temperature:2...8 °C
- MDL No.:MFCD00132688
- Density:0,5 g/ml at 25 °C (lit.)
- Synonym(s):Hydroxypropyl cellulose
- CAS No.:9004-64-2



