Order Entry
Belgium
ContactUsLinkComponent
Pharmaceutical Analytical Standard, Escitalopram for System Suitability, Reference Material
Pharmaceutical Analytical Standard, Escitalopram for System Suitability, Reference Material
Catalog # EDQMY0001797
Supplier:  EDQM
CAS Number:  
Pharmaceutical Analytical Standard, Escitalopram for System Suitability, Reference Material
Catalog # EDQMY0001797
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    15 mg
  • Standards:
    Analytical reference standard
  • CAS No.:
    219861-08-2
  • Description:
    Escitalopram for system suitability
  • MDL No.:
    MFCD06407826
  • Synonym(s):
    Escitalopram oxalate, S-(+)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate, S-(+)-Citalopram oxalate
  • Molecular weight:
    414,43
  • Storage temperature:
    2...8 °C

Specifications

About this item

Escitalopram for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Escitalopram is a selective serotonin reuptake inhibitor (SSRI), the S-enantiomer and eutomer of citalopram.