Pharmaceutical Analytical Standard, Rhein, Reference Material
Catalog # EDQMY0002159
Supplier: EDQM
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical reference standard
- CAS No.:478-43-3
- Description:Rhein
- MDL No.:MFCD00009618
- Synonym(s):4,5-Dihydroxyanthraquinone-2-carboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid, Cassic acid, Chrysazin 3-carboxylic acid, Monorhein, Rheic acid, Rhubarb yellow
- Melting point:≥300 °C (lit.)
- Molecular weight:284,22
- Storage temperature:2...8 °C
Specifications
About this item
Rhein EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.