Anionic Surfactant, a-Ten - Test Kit
Catalog # WTWA252102
Supplier: WTW
CAS Number:
Specifications
- Pk:25
- Description:Anionic surfactants, a-Ten 02552, 0,05 - 2,00 mg/L MBAS
Specifications
About this item
a-Ten 02552, anionic surfactants cell test - 25 determinations.
- Sample volume: 5 ml
- Cuvette: 16 mm, round
Usable with PhotoLab® 6X00, 7X00, S12 - analogue EPA 425.1, APHA 5540 C, ASTM 2330-02, - anal 1 * 25 items.