ContactUsLinkComponent
Microflex® Mega Texture 93-256 Disposable Nitrile Gloves
Microflex® Mega Texture 93-256 Disposable Nitrile Gloves
Supplier:  Ansell
Certificates
Microflex® Mega Texture 93-256 Disposable Nitrile Gloves
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE93-256/8.5-9
  • ANSE93-256/7.5-8
  • ANSE93-256/6.5-7
  • ANSE93-256/10.5-11
  • ANSE93-256/9.5-10
  • Model:
  • Mega Texture 93-256
  • Mega Texture 93-256
  • Mega Texture 93-256
  • Mega Texture 93-256
  • Mega Texture 93-256
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Fully textured
  • Fully textured
  • Fully textured
  • Fully textured
  • Fully textured
  • Cuff style:
  • Elastic
  • Elastic
  • Elastic
  • Elastic
  • Elastic
  • Cuff length:
  • Standard
  • Standard
  • Standard
  • Standard
  • Standard
  • Finger thickness:
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • Palm thickness:
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • Tensile strength:
  • ≥14 MPa
  • ≥14 MPa
  • ≥14 MPa
  • ≥14 MPa
  • ≥14 MPa
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • ASTM No.:
  • D6319
  • D6319
  • D6319
  • D6319
  • D6319