Impurity Reference Material
Catalog # PROOTRC-C366325-50
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- CAS No.:1356190-17-4
- Description:(11beta)-21-Chloro-9-fluoro-11-hydroxy-16-methylpregna-1,4,16-triene-3,20-dione
- Shipping conditions:RT
- Formula:C₂₂H₂₆ClFO₃
- Storage temperature:−20°C
- Molecular weight (Mw):392,89
- Form:Neat