Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetaldehyde ≥95%

Acetaldehyde ≥95%

1 g
Catalog # PROOTRC-A132600-1G
Acetaldehyde ≥95%
Catalog # PROOTRC-A132600-1G
CAS Number:  75-07-0
Acetaldehyde ≥95%

1 g

CAS Number:  75-07-0
Price
327,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Ethanal
  • CAS Number:75-07-0
  • Formula:CH₃CHO
  • MDL Number:MFCD00006991
  • Molecular Weight:44.05 g/mol
Specifications

Specifications

  • Pk:
    1 g

Specifications