Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Polysorbate 20, liquid

Polysorbate 20, liquid
Certificates
Polysorbate 20, liquid

2 Options Available Below

Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Melting Pt:98,9 °C
  • Density:1,1 g/cm³ (25 °C)
  • Flash Pt:275 °C
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL

  • Septum bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L