Orders LinkContactUsLinkComponent
Multi-Parameter Tester, CombiCheck 20
Multi-Parameter Tester, CombiCheck 20
CombiCheck 20 Muliti-parameter standard solutions, 1 KIT
N° de catalogue 713-4097
Fournisseur:   WTW
Multi-Parameter Tester, CombiCheck 20
N° de catalogue 713-4097
Fournisseur:  WTW
Numéro CAS:  
Multi-Parameter Tester, CombiCheck 20
Multi-Parameter Tester, CombiCheck 20
CombiCheck 20 Muliti-parameter standard solutions, 1 KIT
Numéro CAS:  
Prix
136,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
CombiCheck solutions are ready-to-use multi-parameter standards, Each package contains a standard solution and an addition solution. Both solutions can be used directly without dilution for quality assurance.
  • The standard solution is used to check the accuracy of the results for the complete system: procedure - analytical method - reagents - photometer
  • The addition solution is used to check sample-dependent influences by measuring the recovery rate and to determine the most suitable sample preparation method
The maximum number of determinations which can be made with a CombiCheck standard solution depends on the test set used. With the addition solution 280 determinations are always possible.
Parameters can be checked with their concentrations as follows:
Ammonium: 12,0 mg/L NH₄-N
Chloride: 60 mg/L Cl
COD: 750 mg/L COD
Nitrate: 9,0 mg/L NO₃-N
Phosphate: 8,0 mg/L PO₄-P
Sulphate: 500 mg/L SO₄
Spécifications
Spécifications
  • Pk:
    1 KIT
  • Description:
    CombiCheck 20 Muliti-parameter standard solutions
Spécifications