Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Nitrite Standards
Nitrite Standard Solution
Nitrite Standard Solution
Catalog # 713-2320
Supplier:
WTW
CAS Number:
Expand image
Nitrite Standard Solution
Catalog # 713-2320
Supplier:
WTW
CAS Number:
Specifications
Concentration:
1000 mg/L
Pk:
500 mL
Description:
Nitrite standard solution
Specifications
About this item
Product Details
Product Details