Order Entry
Belgium
Orders LinkContactUsLinkComponent
Iron Standard Solution

Iron Standard Solution

Iron standard solution, 1000 mg/L, 500 mL
Catalog # 713-4083
Supplier:   WTW
Iron Standard Solution
Catalog # 713-4083
Supplier:  WTW
CAS Number:  
Iron Standard Solution
Iron Standard Solution

Iron standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
64,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Iron standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.