Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper Standard Solution

Copper Standard Solution

Copper standard solution, 1000 mg/L, 500 mL
Catalog # 713-4087
Supplier:   WTW
Copper Standard Solution
Catalog # 713-4087
Supplier:  WTW
CAS Number:  
Copper Standard Solution
Copper Standard Solution

Copper standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
64,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Copper standard solution
  • Cat. No.:
    713-4087

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.