Order Entry
Belgium
Orders LinkContactUsLinkComponent
Calcium Standard Solution

Calcium Standard Solution

Calcium standard solution, 1000 mg/L, 500 mL
Catalog # 713-4077
Supplier:   WTW
Calcium Standard Solution
Catalog # 713-4077
Supplier:  WTW
CAS Number:  
Calcium Standard Solution
Calcium Standard Solution

Calcium standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
64,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Calcium standard solution

Specifications