Order Entry
Belgium
ContactUsLinkComponent
Calcein AM 4 mM in DMSO ≥90% (by HPCE), clear solution
Calcein AM 4 mM in DMSO ≥90% (by HPCE), clear solution
Catalog # C1359-100UL
CAS Number:  
undefined
Calcein AM 4 mM in DMSO ≥90% (by HPCE), clear solution
Catalog # C1359-100UL
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Seq:
    1286

Specifications

Specification Test Results

  • Appearance:
    Colorless and White to Off White
  • Appearance (Form):
    Liquid
  • Wavelength
    Lambda Max, before hydrolysis:
    ≤300 nm
  • Purity (HPLC):
    ≥90%

Specification Test Results

About this item

 :
 :994.86 g/mol
CAS Number:148504-34-1
Formula:C₄₆H₄₆N₂O₂₃
MDL Number:MFCD05861516