ContactUsLinkComponent
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Catalog # 734-3635
   Low Manufacturing Impact
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Catalog # 734-3635
   Low Manufacturing Impact
Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Colour:
    Clear
  • Description:
    Single use microplate with lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS
  • Pk:
    1
Specifications
About this item
Experience optimal performance with Nunc™ plates with Nunclon™ Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon™ Supra surface is more hydrophilic than the Nunclon™ Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
  • Consistent lot-to-lot performance
  • Sterile with SAL 10⁻⁶
  • Non-cytotoxic
  • Non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • Five years shelf life
The Nunclon™ Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon™ Supra cultureware is a great xeno-free solution for cell therapy research applications.