Pressure Transducers
Supplier: Spectrum Laboratories
About this item
5 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
- Catalog No:
- SPECACPM-05TC-C1S
- SPECACPM-10TC-01S
- SPECACPM-10TC-C1S
- SPECACPM-10TC-01N
- SPECACPM-10TC-C1N
- Accuracy:
- ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
- ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
- ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
- ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
- ±2% from 0 - 6 psi, ±3% from 6 - 30 psi
Specifications
Product Family Options
Product Information
- RangeAvailabilityPrice
- SPECACPM-05TC-C1SPressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibratedPressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated−10 - +75 psiLog in for availability1 * 25 STSpecifications:
More
Specifications
Cat. No.SPECACPM-05TC-C1SAccuracy±2% from 0 - 6 psi, ±3% from 6 - 30 psi - SPECACPM-10TC-01SPressure Transducer, 1×1" TC, polysulfone, sterileSpecifications:
More
Specifications
Cat. No.SPECACPM-10TC-01SAccuracy±2% from 0 - 6 psi, ±3% from 6 - 30 psi - SPECACPM-10TC-C1SPressure Transducer, 1×1" TC, polysulfone, sterile, calibratedPressure Transducer, 1×1" TC, polysulfone, sterile, calibrated−10 - +75 psiLog in for availability1 * 25 STSpecifications:
More
Specifications
Cat. No.SPECACPM-10TC-C1SAccuracy±2% from 0 - 6 psi, ±3% from 6 - 30 psi - SPECACPM-10TC-01NPressure transducer, 1×1" TC, polysulfone, non sterileSpecifications:
More
Specifications
Cat. No.SPECACPM-10TC-01NAccuracy±2% from 0 - 6 psi, ±3% from 6 - 30 psi - SPECACPM-10TC-C1NPressure Transducer, 1×1" TC, polysulfone, non sterile, calibratedPressure Transducer, 1×1" TC, polysulfone, non sterile, calibrated−10 - +75 psiLog in for availability1 * 1 STSpecifications:
More
Specifications
Cat. No.SPECACPM-10TC-C1NAccuracy±2% from 0 - 6 psi, ±3% from 6 - 30 psi
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



