Order Entry
Belgium
ContactUsLinkComponent
Iodine ≥99.8% (by titrimetric analysis), ReagentPlus®, Supelco®
Iodine ≥99.8% (by titrimetric analysis), ReagentPlus®, Supelco®
Catalog # SUPLI3380-50G
Supplier:  Merck
CAS Number:  
Iodine ≥99.8% (by titrimetric analysis), ReagentPlus®, Supelco®
Catalog # SUPLI3380-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :253.81 g/mol
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355