Order Entry
Belgium
ContactUsLinkComponent
Sodium acetate 99.0-101.0% (by total nitrogen and titration analysis), LiChropur™ suitable for HPLC, Supelco®
Sodium acetate 99.0-101.0% (by total nitrogen and titration analysis), LiChropur™ suitable for HPLC, Supelco®
Catalog # 79714-250G
Supplier:  Merck
CAS Number:  
Sodium acetate 99.0-101.0% (by total nitrogen and titration analysis), LiChropur™ suitable for HPLC, Supelco®
Catalog # 79714-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :82.03 g/mol
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459