Cellulose DS-0, powder suitable for thin layer chromatography (TLC), Supelco®
Catalog # 22197-250G-F
Supplier: Merck
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:342.3 g/mol
CAS Number:9004-34-6
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512