0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Silica gel
Silica gel, FluoroFlash® ~40 μm particle size, Supelco®
Silica gel, FluoroFlash® ~40 μm particle size, Supelco®
08965-100G-F
:
Merck
:
Silica gel, FluoroFlash® ~40 μm particle size, Supelco®
08965-100G-F
:
Merck
:
Pk:
100 g
Seq:
3401
:
:
60.08 g/mol
:
Ambient
:
63231-67-4
:
7631-86-9
:
SiO₂
:
MFCD00011232