Laboratory Glassware Washers, GW4290
: SMEG
The GW4290 90 cm glass washer is equipped with a soft touch colour control panel that allows simple and intuitive daily use.
- Colour touch display
- Detergent compartment
- Maximum loading flexibility
- Data recording
- Complete drying
The compact design of the interface, which gives linearity to the surface, does not neglect the completeness of the information: the operator has at his disposal a three-level range of settings complete with 20 pre-set programmes and 20 fully customisable ones to satisfy every need.
- Catalog No:
- SMEGGW4290-00-000
- SMEGGW4290-S0-000
- SMEGGW4290-SC-000
- Model:
- GW4290-00-000
- GW4290-S0-000
- GW4290-SC-000
- Electrical:
- 3 N AC, 400 V, 50 Hz
- 3 N AC, 400 V, 50 Hz
- 3 N AC, 400 V, 50 Hz
- Noise level:
- <50 dBA
- <50 dBA
- <50 dBA
- Water connection:
- Cold, hot and demineralised water connections
- Cold, hot and demineralised water connections
- Cold, hot and demineralised water connections
- W×D×H:
- 900×600×850 mm
- 900×600×850 mm
- 900×600×850 mm
Frequently Bought Together