Orders LinkContactUsLinkComponent
Resolvex® Column Temperature Controller
Resolvex® Column Temperature Controller
Supplier:   Tecan
Resolvex® Column Temperature Controller
Supplier:  Tecan
Certificates
Resolvex® Column Temperature Controller
3 Options Available Below
Product Details & Documents
Column temperature controllers for heating and cooling.
  • Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
  • Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
  • Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).
Product Family Options
Product Information
  • Availability
    Price
  • TCAT282-0240
    Column heater 150 CERA, 1,5×1,5×10"
    Column heater 150 CERA, 1,5×1,5×10"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • TCAT282-0252
    Column heater 250 CERA, 1,5×1,5×17"
    Column heater 250 CERA, 1,5×1,5×17"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • TCAT282-0300
    Column cooler 250 CERA, 1,5×4,0×17"
    Column cooler 250 CERA, 1,5×4,0×17"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST