Robust Temperature Data Logger, EBI 12-T100
About this item
Discover the EBI 12-T100 from ebro: a robust temperature data logger, ideal for independent routine checks in medicine, pharmaceuticals and production. Sterilisable and with a large measuring range for the optimal quality assurance.
- Wide measuring range, –90 to +150 °C
- High-quality PT1000 sensor (class A)
- Robust IP 68 protection class: waterproof and vapour-tight
- Supplied with calibration certificate
- Sterilisable data logger
The EBI 12-T100 from ebro is a high-performance temperature data logger. Its impressive measuring range of –90 to +150 °C and the high-quality PT1000 sensor (class A) offer maximum precision and enable versatile applications from dry ice to steam sterilisers.
Thanks to its robust IP 68 protection class, it is waterproof and vapour-tight. Each device is supplied with a calibration certificate to ensure maximum accuracy.
Dimensions: 48Ø×24H mm.
Factory calibration certificate: –20, 0, 60 and 134 °C.
: (EG) 1935/2004.
3 Options Available Below
Product Details & Documents
Discover the EBI 12-T100 from ebro: a robust temperature data logger, ideal for independent routine checks in medicine, pharmaceuticals and production. Sterilisable and with a large measuring range for the optimal quality assurance.
- Wide measuring range, –90 to +150 °C
- High-quality PT1000 sensor (class A)
- Robust IP 68 protection class: waterproof and vapour-tight
- Supplied with calibration certificate
- Sterilisable data logger
The EBI 12-T100 from ebro is a high-performance temperature data logger. Its impressive measuring range of –90 to +150 °C and the high-quality PT1000 sensor (class A) offer maximum precision and enable versatile applications from dry ice to steam sterilisers.
Thanks to its robust IP 68 protection class, it is waterproof and vapour-tight. Each device is supplied with a calibration certificate to ensure maximum accuracy.
Dimensions: 48Ø×24H mm.
Factory calibration certificate: –20, 0, 60 and 134 °C.
: (EG) 1935/2004.
Manual Robust Temperature Data Logger, EBI 12-T100, Ebro Manual Temperature and Pressure Data Logger EBI 12-EX.pdf
Specifications for Product Family
Specifications
- Catalog No:
- EBRO1340-6600-0100
- EBRO1340-6600
- EBRO1340-6600-EX
- Model:
- EBI 12-T100-Eyelet
- EBI 12-T100
- EBI 12-T100-EX
- Sensor:
- PT1000, class A
- PT1000, class A
- PT1000, class A
- Protection class:
- IP 68
- IP 68
- IP 68
- Recording intervals:
- 250 ms - 24 hrs.
- 250 ms - 24 hrs.
- 250 ms - 24 hrs.
- Weight:
- 90 g
- 90 g
- 90 g
- Data memory - No. of readings:
- 100320
- 100320
- 100320
- Batteries:
- Lithium battery, 3,6 V
- Lithium, 2× BR1225A, 3 V user-replaceable
- Lithium battery, 3,6 V
Specifications
Product Family Options
Data loggers
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- EBRO1340-6600-0100EBI 12-T100-eyelet, calibration points –20, 0, +60 and +134 °C, Ø 48×24H mmEBI 12-T100-eyelet, calibration points –20, 0, +60 and +134 °C, Ø 48×24H mmT: –90…+150 °C±0,5 °C (–90…–40 °C); ±0,2 °C (–40…0 °C); ±0,1 °C (0…120 °C); ±0,05 °C (120…140 °C); ±0,1 °C (140…150 °C)0,01 °CLog in for availability1 * 1 ST2 450,00 €Specifications:
More
Specifications
Cat. No.EBRO1340-6600-0100ModelEBI 12-T100-EyeletSensorPT1000, class AProtection classIP 68Recording intervals250 ms - 24 hrs.Weight90 gData memory - No. of readings100320BatteriesLithium battery, 3,6 V - EBRO1340-6600EBI 12-T100, high temperature data logger, pharmaceutical, medical and food applications; calibration points: –20, 0, 60 and 134 °CEBI 12-T100, high temperature data logger, pharmaceutical, medical and food applications; calibration points: –20, 0, 60 and 134 °CT: –90…+150 °C±0,5 °C (–90…–40 °C); ±0,2 °C (–40…0 °C); ±0,1 °C (0…120 °C); ±0,05 °C (120…140 °C); ±0,1 °C (140…150 °C)0,01 °CLog in for availability1 * 1 ST2 010,00 €Specifications:
More
Specifications
Cat. No.EBRO1340-6600ModelEBI 12-T100SensorPT1000, class AProtection classIP 68Recording intervals250 ms - 24 hrs.Weight90 gData memory - No. of readings100320BatteriesLithium, 2× BR1225A, 3 V user-replaceable - EBRO1340-6600-EXEBI 12-T100-EX, temperature data logger for hazardous areasEBI 12-T100-EX, temperature data logger for hazardous areasEX-areas: –40…+85 °C; T: –90…+150 °C±0,5 °C (–90…–40 °C); ±0,2 °C (–40…0 °C); ±0,1 °C (0…120 °C); ±0,05 °C (120…140 °C); ±0,1 °C (140…150 °C)0,01 °CLog in for availability1 * 1 ST2 200,00 €Specifications:
More
Specifications
Cat. No.EBRO1340-6600-EXModelEBI 12-T100-EXSensorPT1000, class AProtection classIP 68Recording intervals250 ms - 24 hrs.Weight90 gData memory - No. of readings100320BatteriesLithium battery, 3,6 V
Software and readout station
Product Information
- Catalog #DescriptionAvailabilityPrice
- ebro Starter kit, SK 6100, EBI 11-T210 and IF 300 for food and pharmaceutical application, –30…+140 °CLog in for availability1 * 1 ST1 750,00 €
Complementary Products
Product Information
- Catalog #AccuracyDescriptionRangeResolutionAvailabilityPrice
- EBRO1340-6601-0100±0,3 °C (0…105 °C)
±0,5 °C (–90 …–40 °C)EBI 12-T101, data logger for washer and disinfecter; calibration points –20 °C, 0 °C and +60 °CT: –90…+105 °C0,01 °CLog in for availability1 * 1 ST - 1 650,00 € - EBRO1340-6658±0,1 (0…140 °C)
±20 mbarEBI 12-TP237, temperature and pressure data logger, calibration points 0 °C, +60 °C and +134 °C; 100 mbar and 3100 mbar (bei +25 °C); 3100 mbar (at +134 °C)T: 0…140 °C
P: 1…4000 mbar0,01 °C
1 mbarLog in for availability1 * 1 ST - 2 440,00 € - EBRO1340-6601±0,5 °C (–90…–40 °C); ±0,3 °C (–40…105 °C)EBI 12-T101, Temperature data logger, pharmaceutical, medical and food applications (factory calibration certificate: –20 °C, 0 °C and +60 °C)T: –90…+105 °C0,01 °CLog in for availability1 * 1 ST - 1 790,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ringBattery replacement set EBI 12; 3× battery, 6× O-ring620-1693Log in for availability1 * 1 ST - 194,00 €
Product Information
- Catalog #AvailabilityPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12EBRO1100-0130Log in for availability1 * 1 ST - 277,00 €



