Orders LinkContactUsLinkComponent
epT.I.P.S.® Sets, Pipette Tips
epT.I.P.S.® Sets, Pipette Tips
   Sustainable Options Available
Supplier:   EPPENDORF
epT.I.P.S.® Sets, Pipette Tips
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
epT.I.P.S.® Sets, Pipette Tips
8 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organised pipetting in laboratories.
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S.® Box 2.0 and epT.I.P.S.® Set available in Eppendorf Quality
  • ACT® labelled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.
Certifications: General quality and ISO certificates available.
Specifications for Product Family
Specifications
  • Catalog No:
  • EPPE0030076.290
  • EPPE0030076.303
  • EPPE0030076.311
  • EPPE0030076.320
  • EPPE0030076.338
  • EPPE0030076.346
  • EPPE0030076.354
  • EPPE0030076.362
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Product Information
  • Volume
    Sterile
    Length
    Colour code
    Packed
    Availability
    Price
  • 0,1 - 10 µl
    -
    34 mm
    Dark grey
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 70,90 €
    70,90 €
    1 * 480 ST
       70,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.290
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 0,1 - 20 µl
    -
    40 mm
    Grey
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 70,90 €
    70,90 €
    1 * 480 ST
       70,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.303
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 0,5 - 20 µl
    -
    46 mm
    Light grey
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 65,20 €
    65,20 €
    1 * 480 ST
       65,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.311
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 2 - 200 µl
    -
    53 mm
    Yellow
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 62,70 €
    62,70 €
    1 * 480 ST
       62,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.320
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 20 - 300 µl
    -
    55 mm
    Orange
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 62,70 €
    62,70 €
    1 * 480 ST
       62,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.338
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1000 µl
    -
    71 mm
    Blue
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 66,80 €
    66,80 €
    1 * 480 ST
       66,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.346
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1250 µl
    -
    76 mm
    Green
    480 tips (5 trays × 96 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 87,20 €
    87,20 €
    1 * 480 ST
       87,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.354
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 250 - 2500 µl (0,25 - 2,5 ml)
    -
    115 mm
    Red
    240 tips (5 trays × 48 tips), 1 reusable box
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 61,30 €
    61,30 €
    1 * 240 ST
       61,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030076.362
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.