Orders LinkContactUsLinkComponent
epT.I.P.S.® Singles, Pipette Tips
epT.I.P.S.® Singles, Pipette Tips
   Sustainable Options Available
Supplier:   EPPENDORF
epT.I.P.S.® Singles, Pipette Tips
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
epT.I.P.S.® Singles, Pipette Tips
3 Options Available Below
Product Details & Documents
epT.I.P.S.® Singles high-precision pipette tips offer unparalleled precision in 'Biopur®' and 'Sterile' grades, featuring individually wrapped tips for maximum sterility. Ideal for sensitive and critical lab applications where contamination control is paramount.
  • Eppendorf pipette tip, individually wrapped
  • Available in Biopur and Sterile purity grades
  • For use in medical, pharmaceutical, food industry, molecular biology, and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • ACT® labelled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.
Certifications: Lot-specific certficates for the purity grades Sterile, Biopur® can be downloaded from the Eppendorf website.
Specifications for Product Family
Specifications
  • Catalog No:
  • EPPE0030010.027
  • EPPE0030010.043
  • EPPE0030010.060
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Product Information
  • Volume
    Sterile
    Length
    Colour code
    Packed
    Availability
    Price
  • 0,1 - 20 µl
    +
    40 mm
    Grey
    100 tips, individually packed
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,10 €
    35,10 €
    1 * 100 ST
       35,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030010.027
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 2 - 200 µl
    +
    53 mm
    Yellow
    100 tips, individually packed
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,10 €
    35,10 €
    1 * 100 ST
       35,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030010.043
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1000 µl
    +
    71 mm
    Blue
    100 tips, individually packed
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,10 €
    35,10 €
    1 * 100 ST
       35,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030010.060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact