epT.I.P.S.® Bulk - Pipette Tips
Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
- epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available colour-coded yellow and blue
- epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
- Fine retracted or extended tips simplify work with small volumes
- ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.
How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
: General quality and ISO certificates available.
Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
- epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available colour-coded yellow and blue
- epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
- Fine retracted or extended tips simplify work with small volumes
- ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.
How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
: General quality and ISO certificates available.
- Catalog No:
- 613-3500
- 613-3501
- 613-3502
- 613-3111
- 613-3112
- 613-3113
- 613-3106
- 613-3506
- 613-3508
- EPPE0030000.650
- 613-3507
- 613-4269
- 613-4270
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Product Family Options
- VolumeLengthColour codePacked
- 0,1 - 10 µl34 mmDark grey1000 tips (2 bags×500 tips)1 * 1.000 ST72,70 €
Low Manufacturing Impact Specifications
Cat. No.613-3500Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 0,1 - 20 µl40 mmGrey1000 tips (2 bags×500 tips)1 * 1.000 ST72,70 €
Low Manufacturing Impact Specifications
Cat. No.613-3501Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 0,5 - 20 µl46 mmLight grey1000 tips (2 bags×500 tips)1 * 1.000 ST69,20 €
Low Manufacturing Impact Specifications
Cat. No.613-3502Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 2 - 200 µl53 mmYellow1000 tips (2 bags×500 tips)1 * 1.000 ST44,90 €
Low Manufacturing Impact Specifications
Cat. No.613-3111Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 20 - 300 µl55 mmOrange1000 tips (2 bags×500 tips)1 * 1.000 ST44,90 €
Low Manufacturing Impact Specifications
Cat. No.613-3112Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 50 - 1000 µl71 mmBlue1000 tips (2 bags×500 tips)1 * 1.000 ST44,90 €
Low Manufacturing Impact Specifications
Cat. No.613-3113Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 50 - 1250 µl103 mmDark green1000 tips (4 bags×250 tips)1 * 1.000 ST77,30 €
Low Manufacturing Impact Specifications
Cat. No.613-3106Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 50 - 1250 µl76 mmGreen1000 tips (4 bags×250 tips)1 * 1.000 ST76,30 €
Low Manufacturing Impact Specifications
Cat. No.613-3506Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 100 - 5000 µl120 mmViolet500 tips (5 bags×100 tips)1 * 500 ST61,30 €
Low Manufacturing Impact Specifications
Cat. No.613-3508Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 200 - 5000 µl175 mmViolet300 tips (3 bags×100 tips)1 * 300 ST41,80 €
Low Manufacturing Impact Specifications
Cat. No.EPPE0030000.650Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 250 - 2500 µl115 mmRed500 tips (5 bags×100 tips)1 * 500 ST52,50 €
Low Manufacturing Impact Specifications
Cat. No.613-3507Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 500 - 10000 µl165 mmTurquoise200 tips (2 bags×100 tips)1 * 200 ST62,90 €
Low Manufacturing Impact Specifications
Cat. No.613-4269Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001 - 500 - 10000 µl243 mmTurquoise200 tips (2 bags×100 tips)1 * 200 ST89,80 €
Low Manufacturing Impact Specifications
Cat. No.613-4270Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact