Order Entry
Belgium
Orders LinkContactUsLinkComponent
illustra™ ProbeQuant™ G-50 micro columns

illustra™ ProbeQuant™ G-50 micro columns

illustra™ ProbeQuant™ G-50 Micro Columns, 50
Catalog # 28-9034-08
Supplier:   Cytiva
illustra™ ProbeQuant™ G-50 micro columns
Catalog # 28-9034-08
Supplier:  Cytiva
CAS Number:  
illustra™ ProbeQuant™ G-50 micro columns
illustra™ ProbeQuant™ G-50 micro columns

illustra™ ProbeQuant™ G-50 Micro Columns, 50

CAS Number:  
Price
272,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

ProbeQuant™ G-50 micro columns use spin column chromatography to provide a fast and reliable method for purifying labelled probes from unincorporated labelled nucleotides. In addition, a portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation. The columns are designed for use in a microcentrifuge, come prepacked with Sephadex™ G-50 DNA grade and are pre-equilibrated in STE, which means that they are ready to use. They can accommodate samples ranging in volume from 25 μl to 50 μl.

  • For rapid purification of labeled DNA (20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography
  • Prepacked with Sephadex G-50 DNA Grade (see illustra Sephadex G-50 and G-100 DNA Grade) and pre-equilibrated in STE containing 0.15% Kathon CG/ICP Biocide
  • Requires less than 4 minutes from sample application to collection of purified product
  • Designed for use in a microcentrifuge
  • Tested in nickase, single and double-stranded exonuclease and RNase assays

In addition, a portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation. The columns are designed for use in a microcentrifuge, come prepacked with Sephadex G-50 DNA grade and are pre-equilibrated in STE, which means that they are ready to use. They can accommodate samples ranging in volume from 25 to 50 μl.

Specifications

Specifications

  • Pk:
    50
  • Description:
    illustra™ ProbeQuant™ G-50 Micro Columns
  • Cat. No.:
    28-9034-08
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Description:
    illustra™ ProbeQuant™
  • Sample type:
    PCR and other enzymatic reactions

Specifications