Order Entry
Belgium
ContactUsLinkComponent
illustra™ GFX™ PCR DNA and gel band purification kit
illustra™ GFX™ PCR DNA and gel band purification kit
Supplier:  Cytiva
Certificates
undefined
illustra™ GFX™ PCR DNA and gel band purification kit
Supplier:  Cytiva
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
    Amplified/modified DNA
  • Sample type:
    Gel matrix
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    illustra™ GFX™ PCR DNA and Gel Band Purification Kit

Specifications

Frequently Bought Together