Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Irritant
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Catalog # FFCHFF1940500
Supplier:  FF CHEMICALS
CAS Number:  
Irritant
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Catalog # FFCHFF1940500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589