
Hydrochloric acid 10%
Catalog # FFCHFF20755000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324