Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 0.5 mol/l (0.5 N)
Corrosive
Hydrochloric acid 0.5 mol/l (0.5 N)
Catalog # FFCHFF2235000
Supplier:  FF CHEMICALS
CAS Number:  
Corrosive
Hydrochloric acid 0.5 mol/l (0.5 N)
Catalog # FFCHFF2235000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324