Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite (4% Cl₂) in aqueous solution
Sodium hypochlorite (4% Cl₂) in aqueous solution
Catalog # FFCHFF4280500
Supplier:  FF CHEMICALS
CAS Number:  
Sodium hypochlorite (4% Cl₂) in aqueous solution
Catalog # FFCHFF4280500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :74.44 g/mol
Storage Temperature:Refrigerator
EINECS:231-668-3
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120