Sodium hypochlorite (10% Cl₂) in aqueous solution
Glass bottle, 100 mL
Supplier: FF CHEMICALS
Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # FFCHFF2540100
Supplier: FF CHEMICALS
CAS Number: 7681-52-9
Jump to:
Product Details & Documents
- CAS Number:7681-52-9
- Formula:ClNaO
- MDL Number:MFCD00011120
- Molecular Weight:74.44 g/mol
- Boiling Pt:111 °C (1013 hPa)
- Melting Pt:–30…–20 °C
- Density:1.22…1.25 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:231-668-3
- UN:1791
- ADR:8,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



