Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 5% (w/v) in aqueous solution
Corrosive
Sodium hydroxide 5% (w/v) in aqueous solution
Catalog # 35604.AP
CAS Number:  
Corrosive
Sodium hydroxide 5% (w/v) in aqueous solution
Catalog # 35604.AP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548